accessibility menu, dialog, popup

UserName

CH3NOOOHN

CAS RN 33224-01-0

IUPAC Name: 1-methyl-5-oxo-2-(pyridin-3-yl)pyrrolidine-3-carboxylic acid
Synonyms: 1-methyl-5-oxo-2-(pyridin-3-yl)pyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: DEYLVDCFTICBTB-UHFFFAOYNA-N
SMILES: CN1C(C(CC1=O)C(O)=O)C1=CN=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results