missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone 95.0+%, TCI America™

Catalog No. M1053100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1053100MG 100 mg
M10531G 1 g
M10535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1053100MG Supplier TCI America Supplier No. M1053100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33224-01-0 | C11H12N2O3 | 220.228 g/mol

Chemical Identifiers

CAS 33224-01-0
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD00075394
InChI Key DEYLVDCFTICBTB-WCBMZHEXSA-N
PubChem CID 671220
IUPAC Name (2S,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylic acid
SMILES CN1C(C(CC1=O)C(=O)O)C2=CN=CC=C2

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 100 mg
Synonym trans-4′C-Carboxycotinine
Formula Weight 220.23
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone