missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Hexen-1-ylboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7034912
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5261303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5261303 Supplier Thermo Scientific Chemicals Supplier No. H5261303

trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 42599-18-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074553
InChI Key GWFKSQSXNUNYAC-AATRIKPKSA-N
Synonym e-hexen-1-ylboronic acid, e-hex-1-en-1-ylboronic acid, trans-1-hexen-1-ylboronic acid, boronic acid, b-1-hexen-1-yl, 1e-hex-1-en-1-ylboronic acid, boronic acid,b-1e-1-hexenyl, e-hex-1-enylboronic acid, e-1-hexenyldihydroxyborane, hex-1-en-1-ylboronic acid
PubChem CID 5326225
IUPAC Name [(E)-hex-1-enyl]boronic acid
SMILES B(C=CCCCC)(O)O

Specifications

Melting Point 62°C to 63°C
Quantity 1 g
Formula Weight 127.98
Percent Purity 98%
Chemical Name or Material trans-1-Hexen-1-ylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only