missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1-(Boc-amino)-4-ethynylcyclohexane, 97%
CAS: 947141-86-8 | C13H21NO2 | 223.32 g/mol
Supplier: Thermo Scientific Chemicals H6290503
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesIncompatible with oxidizing agents.
Chemical Identifiers
| 947141-86-8 | |
| 223.32 | |
| NZTBBJYAWNVCOE-UHFFFAOYSA-N | |
| 57538360 | |
| CC(C)(C)OC(=O)NC1CCC(CC1)C#C |
| C13H21NO2 | |
| MFCD12963985 | |
| tert-butyl trans-4-ethynylcyclohexylcarbamate, trans-1-boc-amino-4-ethynylcyclohexane, tert-butyl n-4-ethynylcyclohexyl carbamate, nztbbjyawnvcoe-xypyzodxsa-n, tert-butyl trans 4-ethynylcyclohexylcarbamate, tert-butyl trans-4-ethynylcyclohexanecarbamate, tert-butyl trans-4-ethynylcyclohexyl carbamate, trans-4-ethynyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl n-1r,4r-4-ethynylcyclohexyl carbamate, 1alpha,4beta-4-ethynylcyclohexyl carbamic acid tert-butyl ester | |
| tert-butyl N-(4-ethynylcyclohexyl)carbamate |
Specifications
| trans-1-(Boc-amino)-4-ethynylcyclohexane | |
| C13H21NO2 | |
| tert-butyl trans-4-ethynylcyclohexylcarbamate, trans-1-boc-amino-4-ethynylcyclohexane, tert-butyl n-4-ethynylcyclohexyl carbamate, nztbbjyawnvcoe-xypyzodxsa-n, tert-butyl trans 4-ethynylcyclohexylcarbamate, tert-butyl trans-4-ethynylcyclohexanecarbamate, tert-butyl trans-4-ethynylcyclohexyl carbamate, trans-4-ethynyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl n-1r,4r-4-ethynylcyclohexyl carbamate, 1alpha,4beta-4-ethynylcyclohexyl carbamic acid tert-butyl ester | |
| CC(C)(C)OC(=O)NC1CCC(CC1)C#C | |
| 1 g | |
| 57538360 | |
| 97% |
| 947141-86-8 | |
| MFCD12963985 | |
| NZTBBJYAWNVCOE-UHFFFAOYSA-N | |
| tert-butyl N-(4-ethynylcyclohexyl)carbamate | |
| 223.32 | |
| 223.31 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures