accessibility menu, dialog, popup

UserName

CAS RN 947141-86-8

IUPAC Name: tert-butyl N-(4-ethynylcyclohexyl)carbamate
Synonyms: tert-butyl N-(4-ethynylcyclohexyl)carbamate
Molecular Weight (g/mol): 223.32
Molecular Formula: C13H21NO2
InChi Key: NZTBBJYAWNVCOE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC1)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)