missing translation for 'onlineSavingsMsg'
Learn More

trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%

Catalog No. AAH62670MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62670MD 250 mg
AAH6267003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62670MD Supplier Thermo Scientific Chemicals Supplier No. H62670MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1212405-03-2 | C12H22BrNO2 | 292.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1212405-03-2
Molecular Formula C12H22BrNO2
Molecular Weight (g/mol) 292.217
MDL Number MFCD12022626
InChI Key XJDUGNDNCQMMBK-NXEZZACHSA-N
Synonym tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate
PubChem CID 46172881
IUPAC Name tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CBr

Specifications

Quantity 250 mg
Formula Weight 292.21
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only