CAS RN 1212405-03-2
IUPAC Name:
tert-butyl N-[3-(bromomethyl)cyclohexyl]carbamate
Synonyms:
tert-butyl N-[3-(bromomethyl)cyclohexyl]carbamate
Molecular Weight (g/mol):
292.22
Molecular Formula:
C12H22BrNO2
InChi Key:
XJDUGNDNCQMMBK-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC1CCCC(CBr)C1
1
–
1
of
1
results
1
Promotions Available
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%
CAS: 1212405-03-2 Molecular Formula: C12H22BrNO2 Molecular Weight (g/mol): 292.217 MDL Number: MFCD12022626 InChI Key: XJDUGNDNCQMMBK-NXEZZACHSA-N Synonym: tert-butyl trans-3-bromomethyl-cyclohexylcarbamate,t-butyl trans-3-bromomethyl cyclohexyl carbamate,tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate PubChem CID: 46172881 IUPAC Name: tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)CBr