missing translation for 'onlineSavingsMsg'
Learn More

trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%

CAS: 1212405-03-2 | C12H22BrNO2 | 292.217 g/mol

Supplier:  Thermo Scientific Chemicals H6267003

Quantity 1 g

 View more versions of this product

Catalog No. AAH6267003


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1212405-03-2
292.217
XJDUGNDNCQMMBK-NXEZZACHSA-N
46172881
CC(C)(C)OC(=O)NC1CCCC(C1)CBr
C12H22BrNO2
MFCD12022626
tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate
tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate

Specifications

trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane
C12H22BrNO2
tert-butyl trans-3-bromomethyl-cyclohexylcarbamate, t-butyl trans-3-bromomethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-bromomethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCCC(C1)CBr
1 g
46172881
97%
1212405-03-2
MFCD12022626
XJDUGNDNCQMMBK-NXEZZACHSA-N
tert-butyl N-[(1R,3R)-3-(bromomethyl)cyclohexyl]carbamate
292.217
292.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions