missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Pentadiene (stabilized with TBC) 93.0+%, TCI America™

Catalog No. p-7134714
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06471ML 1 mL
P06475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06471ML Supplier TCI America Supplier No. P06471ML
Only null left

Chemical Identifiers

CAS 2004-70-8
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00009295
InChI Key PMJHHCWVYXUKFD-SNAWJCMRSA-N
Synonym trans-Piperylene
PubChem CID 62204
ChEBI CHEBI:74165
IUPAC Name (3E)-penta-1,3-diene
SMILES CC=CC=C

Specifications

Color Colorless
Boiling Point 42°C
Quantity 1 mL
UN Number 3295
Formula Weight 68.12
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,3-Pentadiene (stabilized with TBC)
EINECSNumber (2)-0020
RTECSNumber RZ2465000
TSCA Yes
Recommended Storage Refrigerator