accessibility menu, dialog, popup

UserName

CH3CH2(E)

CAS RN 2004-70-8

IUPAC Name: (3E)-penta-1,3-diene
Synonyms: piperylene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: PMJHHCWVYXUKFD-SNAWJCMRSA-N
SMILES: C\C=C\C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results