missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Dichloropropene 98.0+%, TCI America™

$260.97 - $782.90

Chemical Identifiers

CAS 10061-02-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-OWOJBTEDSA-N
Synonym 1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
PubChem CID 24726
ChEBI CHEBI:18624
IUPAC Name (1E)-1,3-dichloroprop-1-ene
SMILES ClC\C=C\Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D23465ML
View Documents
TCI America
D23465ML
5 mL
Each for $260.97
Only null left
 
D234625ML
View Documents
TCI America
D234625ML
25 mL
Each for $782.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10061-02-6
110.97
UOORRWUZONOOLO-OWOJBTEDSA-N
24726
(1E)-1,3-dichloroprop-1-ene
C3H4Cl2
MFCD00000985
1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
CHEBI:18624
ClC\C=C\Cl

Specifications

10061-02-6
112°C
MFCD00000985
2047
UOORRWUZONOOLO-OWOJBTEDSA-N
(1E)-1,3-dichloroprop-1-ene
24726
110.97
Liquid
Yellow
C3H4Cl2
5 mL
1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
ClC\C=C\Cl
110.97
CHEBI:18624
≥98.0% (GC)
trans-1,3-Dichloropropene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0125

RTECSNumber : UC8320000

TSCA : Yes

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'