missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Dichloropropene 98.0+%, TCI America™

Catalog No. D23465ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23465ML 5 mL
D234625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23465ML Supplier TCI America Supplier No. D23465ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10061-02-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-OWOJBTEDSA-N
Synonym 1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
PubChem CID 24726
ChEBI CHEBI:18624
IUPAC Name (1E)-1,3-dichloroprop-1-ene
SMILES ClC\C=C\Cl

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 mL
UN Number 2047
Formula Weight 110.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,3-Dichloropropene
EINECSNumber (2)-0125
RTECSNumber UC8320000
TSCA Yes
Recommended Storage Freezer