missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichlorocyclohexane 96.0+%, TCI America™

Catalog No. D22305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22305G 5 g
D223025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22305G Supplier TCI America Supplier No. D22305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 822-86-6
Molecular Formula C6H10Cl2
Molecular Weight (g/mol) 153.05
MDL Number MFCD00003824
InChI Key GZEZIBFVJYNETN-UHFFFAOYNA-N
Synonym trans-1,2-dichlorocyclohexane, 1r,2r-1,2-dichlorocyclohexane, t-1,2-dichlorocyclohexane, cyclohexane, 1,2-dichloro-, 1r,2r-rel, trans-1,2-dichlorcyclohexan, 1beta,2alpha-dichlorocyclohexane, gzezibfvjynetn-phdidxhhsa, 1,2-dichlorocyclohexane, trans
PubChem CID 2723623
IUPAC Name 1,2-dichlorocyclohexane
SMILES ClC1CCCCC1Cl

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 g
Formula Weight 153.05
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,2-Dichlorocyclohexane