accessibility menu, dialog, popup

UserName

ClCl

CAS RN 822-86-6

IUPAC Name: 1,2-dichlorocyclohexane
Synonyms: 1,2-dichlorocyclohexane -trans 1,2-dichlorocyclohexane -cis
Molecular Weight (g/mol): 153.05
Molecular Formula: C6H10Cl2
InChi Key: GZEZIBFVJYNETN-UHFFFAOYNA-N
SMILES: ClC1CCCCC1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results