missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibenzoylethylene 98.0+%, TCI America™

Catalog No. D21855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21855G 5 g
D218525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21855G Supplier TCI America Supplier No. D21855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 959-28-4
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00003083
InChI Key WYCXGQSQHAXLPK-VAWYXSNFSA-N
Synonym trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
PubChem CID 735960
IUPAC Name (E)-1,4-diphenylbut-2-ene-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 110°C
Color Yellow
Quantity 5 g
Formula Weight 236.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Dibenzoylethylene
RTECSNumber EM7000000
TSCA Yes