missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%

Catalog No. p-3736120
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC155000250 25 g
AC155005000 500 g
AC155000025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155000250 Supplier Thermo Scientific Chemicals Supplier No. 155000250
Only null left

trans-1,2-Diaminocyclohexane-N,N,N,N-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4 | CAS: 125572-95-4 | C14H20N2O8 | 344.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym 2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
PubChem CID 2723844
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Melting Point >210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Complexometry)
Packaging Plastic Bottle
Linear Formula C6H10[N(CH2CO2H)2]2·H2O
Quantity 25 g
Beilstein 13,III,1
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common organic solvents, soluble in alkali solutions
IUPAC Name 2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
Formula Weight 364.35
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only