Learn More
(+/-)-trans-1,2-Diaminocyclohexane, 99%
CAS: 1121-22-8 | C6H14N2 | 114.192 g/mol
Supplier: Thermo Scientific Chemicals L0662618
Description
(±)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(^+)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
| (+/-)-trans-1,2-Diaminocyclohexane | |
| 14°C to 15°C | |
| 89°C to 90°C (14 mmHg) | |
| Amine-like | |
| 1.489 | |
| 50 g | |
| 3193807 | |
| 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine | |
| SSJXIUAHEKJCMH-PHDIDXHHSA-N | |
| (1R,2R)-cyclohexane-1,2-diamine | |
| 43806 | |
| 99% |
| 1121-22-8 | |
| 0.951 | |
| 68°C (154°F) | |
| C6H14N2 | |
| MFCD00063747 | |
| UN2735 | |
| Air sensitive; May form a precipitate | |
| Miscible with water. | |
| C1CCC(C(C1)N)N | |
| 114.192 | |
| 114.19 |
Chemical Identifiers
| 1121-22-8 | |
| 114.192 | |
| SSJXIUAHEKJCMH-PHDIDXHHSA-N | |
| 43806 | |
| C1CCC(C(C1)N)N |
| C6H14N2 | |
| MFCD00063747 | |
| 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine | |
| (1R,2R)-cyclohexane-1,2-diamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only