missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-trans-1,2-Diaminocyclohexane, 99%

CAS: 1121-22-8 | C6H14N2 | 114.192 g/mol

$284.22 - $937.38

Identifiants chimiques

CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAL0662618
Afficher les documents
Thermo Scientific Chemicals
L0662618
50 g
chaque for $284.22
Il en reste null
 
AAL0662630
Afficher les documents
Thermo Scientific Chemicals
L0662630
250 g
chaque for $937.38
Il en reste null
 
Description

Description

(±)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Spécifications

Identifiants chimiques

1121-22-8
114.192
SSJXIUAHEKJCMH-PHDIDXHHSA-N
43806
C1CCC(C(C1)N)N
C6H14N2
MFCD00063747
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
(1R,2R)-cyclohexane-1,2-diamine

Spécifications

1121-22-8
0.951
68°C (154°F)
C6H14N2
MFCD00063747
UN2735
Air sensitive; May form a precipitate
Miscible with water.
C1CCC(C(C1)N)N
114.192
114.19
(+/-)-trans-1,2-Diaminocyclohexane
14°C to 15°C
89°C to 90°C (14 mmHg)
Amine-like
1.489
50 g
3193807
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
SSJXIUAHEKJCMH-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diamine
43806
99%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Nitrogen

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only