missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanediol 99.0+%, TCI America™
$37.29 - $177.08
Chemical Identifiers
| CAS | 1460-57-7 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00063611 |
| InChI Key | PFURGBBHAOXLIO-PHDIDXHHSA-N |
| Synonym | trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol |
| PubChem CID | 92886 |
| ChEBI | CHEBI:16931 |
| IUPAC Name | (1R,2R)-cyclohexane-1,2-diol |
| SMILES | O[C@@H]1CCCC[C@H]1O |
Chemical Identifiers
| 1460-57-7 | |
| 116.16 | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| 92886 | |
| (1R,2R)-cyclohexane-1,2-diol |
| C6H12O2 | |
| MFCD00063611 | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| CHEBI:16931 | |
| O[C@@H]1CCCC[C@H]1O |
Specifications
| 1460-57-7 | |
| White | |
| C6H12O2 | |
| 5 g | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| (1R,2R)-cyclohexane-1,2-diol | |
| 92886 | |
| 116.16 | |
| Crystalline Powder |
| 102°C | |
| 120°C | |
| MFCD00063611 | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| O[C@@H]1CCCC[C@H]1O | |
| 116.16 | |
| CHEBI:16931 | |
| ≥99.0% (GC) | |
| trans-1,2-Cyclohexanediol |
Safety and Handling
TSCA : No