missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanediol 99.0+%, TCI America™
Supplier: TCI America C080925G
Specifications
| trans-1,2-Cyclohexanediol | |
| 102°C | |
| 120°C | |
| MFCD00063611 | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| O[C@@H]1CCCC[C@H]1O | |
| 116.16 | |
| CHEBI:16931 | |
| ≥99.0% (GC) |
| 1460-57-7 | |
| White | |
| C6H12O2 | |
| 25 g | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| (1R,2R)-cyclohexane-1,2-diol | |
| 92886 | |
| 116.16 | |
| Crystalline Powder |
Chemical Identifiers
| 1460-57-7 | |
| 116.16 | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| 92886 | |
| (1R,2R)-cyclohexane-1,2-diol |
| C6H12O2 | |
| MFCD00063611 | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| CHEBI:16931 | |
| O[C@@H]1CCCC[C@H]1O |
Safety and Handling
TSCA : No