missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediaminetetraacetic Acid Monohydrate 99.0+%, TCI America™

Catalog No. C047325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C047325G 25 g
C0473100G 100 g
C0473500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C047325G Supplier TCI America Supplier No. C047325G

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
Synonym 2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
PubChem CID 2723844
IUPAC Name 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

Melting Point 216°C
Color White
Quantity 25 g
Formula Weight 346.34
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Cyclohexanediaminetetraacetic Acid Monohydrate
RTECSNumber AG5000000
TSCA Yes