missing translation for 'onlineSavingsMsg'
Learn More

Tranilast 98.0+%, TCI America™

Catalog No. T29351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29351G 1 g
T29355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29351G Supplier TCI America Supplier No. T29351G

Chemical Identifiers

CAS 53902-12-8
Molecular Formula C18H17NO5
Molecular Weight (g/mol) 327.34
MDL Number MFCD00864787
InChI Key NZHGWWWHIYHZNX-CSKARUKUSA-N
Synonym tranilast, rizaben, trans-tranilast, tranilastum, n-3,4-dimethoxycinnamoyl anthranilic acid, tranilastum inn-latin, tranilast usan:inn:jan, tranpro, unii-hvf50smy6e, rizaben tn
PubChem CID 5282230
ChEBI CHEBI:77572
IUPAC Name 2-[(2E)-3-(3,4-dimethoxyphenyl)prop-2-enamido]benzoic acid
SMILES COC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1OC

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 327.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tranilast
RTECSNumber DG8731000
TSCA No
Recommended Storage Refrigerator