missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012735
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012735 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012735 Supplier Merck Emd Millipore Supplier No. PHR1812500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Pharmaceutical (small molecule)

Specifications

CAS 1197-18-8
Linear Formula H2 NCH2C6H10 CO2 H
MDL Number MFCD00001466
Melting Point >300°C
Synonym trans-4-(Aminomethyl)cyclohexanecarboxylic acid; AMCA; AMCHA; HAKU; TAMCHA; Tranexamic acid
Type Certified Reference Material
Quantity 500 mg
Formula Weight 157.21
Grade Certified Reference Material