Learn More
Eppendorf™ Trade-In Research™ Plus
Trade-In Research Plus pipettes do not have to be in working condition and may be any manufacturers brand.
Supplier: Eppendorf™ 2231300029
Description
- Returned pipettes do not have to be in working condition and may be any manufacturers brand.
- Pipettes must be free of contamination and returned together with the signed Decontamination Form (www.eppendorf.com/decontamination) to the following address:
- In U.S.: Eppendorf Services, 151 Memorial Drive, Suite B, Shrewsbury, MA 01545
- In Canada: Eppendorf Canada, 2810 Argentia Road, Unit #2, Mississauga, ONT L5N 8L2
- If you have any questions, please contact Customer Support at:
- U.S.: 800-645-3050
- Canada: 800-263-8715
- Benefit from one of the lowest operating and tip ejection forces on the pipette market, helping to reduce hand and arm strain
- Temporarily adjust your pipette in seconds for better accuracy when pipetting challenging liquids like ethanol or when pipetting at high altitudes. Return to factory adjustment without calibration
- Autoclave the entire pipette or only the lower part according to your needs and applications to ensure a proper decontamination
- Premium material ensures a high UV and chemical resistance
- Enjoy excellent flexibility and choose among single-channel pipettes in fixed or variable volume as well as 8-, 12-, 16- and 24-channel pipettes
Specifications
| Autoclavable | |
| Pipette | |
| Blue |
| 1 | |
| 200 μL |