missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Trans-Caffeic Acid, MilliporeSigma™ Supelco™

Catalog No. 111014342
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014342 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014342 Supplier Merck Emd Millipore Supplier No. 5186850MG
Only null left
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 501-16-6
Product Line TraceCERT
Physical Form Neat
Linear Formula C9H8O4
MDL Number MFCD00004392
Synonym trans-3,4-Dihydroxycinnamic acid
Type Certified Reference Material
Quantity 50 mg
Formula Weight 180.16
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC