missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Taurine, MilliporeSigma™ Supelco™

Catalog No. 111012411
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012411 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012411 Supplier Merck Emd Millipore Supplier No. 93019100MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
CAS 107-35-7
Linear Formula NH2 CH2 CH2 SO3 H
MDL Number MFCD00008197
Melting Point >300°C
Synonym 2-Aminoethanesulfonic acid
Type Certified Reference Material
Quantity 100 mg
Formula Weight 125.15
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC