missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Supelco 37 Component FAME Mix in Dichloromethane (Varied Conc.), MilliporeSigma™ Supelco™

Catalog No. 111017832
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017832 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017832 Supplier Merck Emd Millipore Supplier No. CRM47885

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Components: 
Methyl butyrate 400 μg/mL; Methyl hexanoate 400 μg/mL; Methyl octanoate 400 μg/mL; Methyl decanoate 400 μg/mL; Methyl undecanoate 200 μg/mL; Methyl laurate 400 μg/mL; Methyl tridecanoate 200 μg/mL; Methyl myristate 400 μg/mL; Methyl myristoleate 200 μg/mL; Methyl pentadecanoate 200 μg/mL; Methyl cis-10-pentadecenoate 200 μg/mL; Methyl palmitate 600 μg/mL; Methyl palmitoleate 200 μg/mL; Methyl heptadecanoate 200 μg/mL; cis-10-Heptadecenoic acid methyl ester 200 μg/mL; Methyl stearate 400 μg/mL; trans-9-Elaidic acid methyl ester 200 μg/mL; cis-9-Oleic acid methyl ester 400 μg/mL; Methyl linolelaidate 200 μg/mL; Methyl linoleate 200 μg/mL; Methyl arachidate 400 μg/mL; Methyl γ-linolenate 200 μg/mL; Methyl cis-11-eicosenoate ≤ 200 μg/mL; Methyl linolenate 200 μg/mL; Methyl heneicosanoate 200 μg/mL; cis-11,14-Eicosadienoic acid methyl ester 200 μg/mL; Methyl behenate 400 μg/mL; cis-8,11,14-Eicosatrienoic acid methyl ester 200 μg/mL; Methyl erucate 200 μg/mL; cis-11,14,17-Eicosatrienoic acid methyl ester 200 μg/mL; cis-5,8,11,14-Eicosatetraenoic acid methyl ester 200 μg/mL; Methyl tricosanoate 200 μg/mL; cis-13,16-Docosadienoic acid methyl ester 200 μg/mL; Methyl lignocerate 400 μg/mL; cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester 200 μg/mL; Methyl nervonate 200 μg/mL; cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 200 μg/mL.

Specifications

Packaging Ampule
CAS 75-09-2
Product Line TraceCERT
Shipping Condition Dry Ice
Physical Form Liquid
Concentration Varied Conc.
Type Certified Reference Material
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC