missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Sulfate Standard for IC, 1,000 mg/L Sulfate in Water, MilliporeSigma™ Supelco™

Catalog No. 111016053
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016053 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016053 Supplier Merck Emd Millipore Supplier No. 90071100ML

missing translation for 'validMainframeMsg'

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label.

Specifications

Product Line TraceCERT
Purity Grade Notes 1000 mg/L sulfate aq. soln., prepared with high purity Na2 SO4 and water
Concentration 1000 mg/L sulfate aq. soln.
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC