missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Sulfadiazine, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 68-35-9 |
| Linear Formula | C10H10N4O2 S |
| MDL Number | MFCD00006065 |
| Melting Point | 253°C |
| Synonym | 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide; N1-(Pyrimidin-2-yl)sulfanilamide |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 250.28 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC