missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Silicon Standard for AAS, 1,000 mg/L Si in NaOH, MilliporeSigma™ Supelco™

Catalog No. 111016688
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016688 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016688 Supplier Merck Emd Millipore Supplier No. 16259250ML
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.Certified content incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 1310-73-2
Product Line TraceCERT
Purity Grade Notes 1000 mg/L Si in 2% sodium hydroxide, prepared with high purity silicon metal, NaOH and water
Concentration 1000 mg/L Si in NaOH
Type Certified Reference Material
Quantity 250 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC