missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Scandium Standard for ICP, 10,000 mg/L Sc in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111016832
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016832 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016832 Supplier Merck Emd Millipore Supplier No. 92504100ML
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.

Specifications

CAS 7697-37-2
Product Line TraceCERT
Purity Grade Notes 10,000 mg/L Sc in 5% nitric acid, prepared with high purity Sc2O3 and HNO3
Concentration 10,000 mg/L Sc in nitric acid
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC