missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Pyrazophos, MilliporeSigma™ Supelco™

Catalog No. 111002348
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002348 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002348 Supplier Merck Emd Millipore Supplier No. 9060250MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Product Line TraceCERT
CAS 13457-18-6
Linear Formula C14H20N3O5 PS
MDL Number MFCD00055494
Type Certified Reference Material
Quantity 50 mg
Formula Weight 373.36
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC