missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Propionaldehyde-DNPH Solution, 1,000 μg/mL in Acetonitrile (as the aldehyde), MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 725-00-8 |
| Flash Point | 2.0°C - Closed Cup |
| Synonym | Propionaldehyde-2,4-dinitrophenylhydrazone solution |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL in acetonitrile (as the aldehyde) |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC