missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Potassium sorbate, MilliporeSigma™ Supelco™

Catalog No. 111013683
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013683 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013683 Supplier Merck Emd Millipore Supplier No. 40430100MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Product Line TraceCERT
CAS 24634-61-5
Physical Form Neat
Linear Formula CH3 CH=CHCH=CHCOOK
MDL Number MFCD00016546
Synonym Potassium 2,4-hexadienoate; Sorbic acid potassium salt
Type Certified Reference Material
Quantity 100 mg
Formula Weight 150.22
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC