missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Potassium Phthalate Monobasic Standard, MilliporeSigma™ Supelco™

Catalog No. 111015731
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015731 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015731 Supplier Merck Emd Millipore Supplier No. 146591G
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Description qNMR Standard for D2O (8.2 ppm/7.5 ppm)
CAS 877-24-7
Product Line TraceCERT
Physical Form Neat
Linear Formula HOOCC6H4 COOK
MDL Number MFCD00013070
pH 4
Melting Point 295°C to 300°C
Synonym KHP; Phthalic acid monopotassium salt; Potassium biphthalate; Potassium hydrogen phthalate
Type Certified Reference Material
Quantity 1 g
Formula Weight 204.22
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC