missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ β-Pinene, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017297
Change view
Click to view available options
Quantity:
2000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017297 2000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017297 Supplier Merck Emd Millipore Supplier No. CRM40433

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 127-91-3
Product Line TraceCERT
Flash Point 9.7°C - closed cup
Concentration 2,000 μg/mL in methanol
Type Certified Reference Material
Quantity 2000 μg
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC