missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ α-Pinene, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017811
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017811 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017811 Supplier Merck Emd Millipore Supplier No. CRM40339

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 80-56-8
Product Line TraceCERT
Flash Point 9.7°C - closed cup
Concentration 2,000 μg/mL in methanol
Type Pharmaceuticals, Illicit Drugs and Alcohol Standards
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC