missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Phosphorus Standard for ICP, 10,000 mg/L P in Water, MilliporeSigma™ Supelco™

Catalog No. 111002538
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002538 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002538 Supplier Merck Emd Millipore Supplier No. 19916100ML

Specifications

Product Line TraceCERT
CAS 7664-38-2
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 10,000 mg/L P aq. soln.

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC