missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Phenol Solution, 500 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020555
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020555 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020555 Supplier Merck Emd Millipore Supplier No. 48688
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 108-95-2
Linear Formula C6H6 O
MDL Number MFCD00002143
Type Food and Cosmetic Component Standard
Quantity 1 mL
Formula Weight 94.11
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 500 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC