missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Phenanthrene-d10 Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111017892
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017892 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017892 Supplier Merck Emd Millipore Supplier No. 48710U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 1517-22-2
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in dichloromethane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC