missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Perylene Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™

Catalog No. 111020267
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020267 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020267 Supplier Merck Emd Millipore Supplier No. 48079

Perylene is a carcinogenic compound, belonging to the class of polycyclic aromatic hydrocarbons, widely distributed in the environment from different sources.

Environmental

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 198-55-0
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in dichloromethane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC