missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Paclobutrazol certified reference material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 76738-62-0 |
| Linear Formula | C15H20 ClN3 O |
| MDL Number | MFCD01678673 |
| Synonym | alpha-tert-Butyl-beta-(4-chlorobenzyl)-1 H-1,2,4-triazole-1-ethanol |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 293.79 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC