missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Naphthalene certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111019059
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019059 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019059 Supplier Merck Emd Millipore Supplier No. 91489100MG
Only null left

Naphthalene is an aromatic hydrocarbon, found to be nonpolar in nature. It is generally produced from petroleum or coal-tar.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
Ignition Point 978°F
CAS 91-20-3
Vapor Pressure 0.03 mmHg (25°C)
Vapor Density 4.4 (vs. air)
Flash Point 80°C
Physical Form Neat
Linear Formula C10H8
MDL Number MFCD00001742
Melting Point 80°C to 82°C
Boiling Point 218°C
Type Certified Reference Material
Quantity 100 mg
Formula Weight 128.17
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC