missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ N-Nitrosodimethylamine Solution, 200 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020562
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020562 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020562 Supplier Merck Emd Millipore Supplier No. 48670

N-Nitrosodimethylamine is a toxic type of N-nitrosamine, which can be commonly synthesized in vivo by intestinal microbiota type of microorganisms. It can undergo reduction to form dimethylamine using zero-valent metal ions like Fe and Zn in the presence of Cu(II) ions.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 62-75-9
Type Food and Cosmetic Component Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 200 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC