missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Multielement Standard Solution 5 for ICP, 10% Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111016877
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016877 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016877 Supplier Merck Emd Millipore Supplier No. 54704100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.Certified content incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 7697-37-2
Product Line TraceCERT
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Cr, 10 mg/L

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC