missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Multi Cation Standard 2 for IC, 0.1% in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111004005
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004005 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004005 Supplier Merck Emd Millipore Supplier No. 93159100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Line TraceCERT
Purity Grade Notes For the preparation of this CRM only materials of highest purity were used.
Concentration ≤0.1% in nitric acid
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) NH 4 , 100 mg/L

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC