missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Methylparaben, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Supplier: Merck Emd Millipore 7972150MG
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| TraceCERT | |
| HOC6H4 CO2 CH3 | |
| 125°C to 128°C | |
| Certified Reference Material | |
| 152.15 |
| 99-76-3 | |
| MFCD00002352 | |
| Methyl 4-hydroxybenzoate; p-Hydroxybenzoic acid methyl ester; Methyl paraben; NIPAGIN | |
| 50 mg | |
| TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC