missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Methomyl, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022768
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022768 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022768 Supplier Merck Emd Millipore Supplier No. 7408850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 16752-77-5
Molecular Formula C5H10N2O2S
Linear Formula C5H10N2O2S
MDL Number MFCD00055451
Quantity 50 mg
Formula Weight 162.21
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC