missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Malonic Acid, MilliporeSigma™ Supelco™

Catalog No. 111013084
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013084 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013084 Supplier Merck Emd Millipore Supplier No. 68714100MG
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 141-82-2
Product Line TraceCERT
Flash Point 157°C
Physical Form Neat
Linear Formula CH2(COOH)2
MDL Number MFCD00002707
Melting Point 132°C to 135°C
Synonym Propanedioic acid
Type Certified Reference Material
Quantity 100 mg
Formula Weight 104.06
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC