missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Magnesium Standard for ICP, 1,000 mg/L Mg in Nitric Acid, MilliporeSigma™ Supelco™

Catalog No. 111016795
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016795 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016795 Supplier Merck Emd Millipore Supplier No. 30083100MLF
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM. Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.

Specifications

CAS 7697-37-2
Product Line TraceCERT
Purity Grade Notes 1000 mg/L Mg in 2% nitric acid, prepared with high purity Mg(NO3)2, HNO3 and water
Concentration 1000 mg/L Mg in nitric acid
Type Certified Reference Material
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC